missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-tert-butyl phosphite, 95%

As a solvent, as an antioxidant, and as an intermediate

Manufacturer:  Alfa Aesar™ 3023303

 View more versions of this product

Catalog No. AA3023303



Specifications

Specifications

Di-tert-butyl phosphite
66°C to 68°C (0.5mmHg)
13086-84-5
1.4200
C8H18O3P-
Phosphorous acid di-tert-butyl ester
HPRZDGKQDIMUTG-UHFFFAOYSA-N
ditert-butyl phosphite
21959072
Liquid
0.995g/mL
1g
Air & Moisture Sensitive
95%
MFCD00014999
Soluble in organic solvents
CC(C)(C)OP([O-])OC(C)(C)C
193.203
194.21

Chemical Identifiers

13086-84-5
193.203
HPRZDGKQDIMUTG-UHFFFAOYSA-N
21959072
CC(C)(C)OP([O-])OC(C)(C)C
C8H18O3P-
MFCD00014999
Phosphorous acid di-tert-butyl ester
ditert-butyl phosphite
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents