Learn More
Di-tert-butyl phosphite, 95%, Thermo Scientific Chemicals
As a solvent, as an antioxidant, and as an intermediate
$61.73 - $463.66
Chemical Identifiers
CAS | 13086-84-5 |
---|---|
Molecular Formula | C8H18O3P |
Molecular Weight (g/mol) | 193.20 |
MDL Number | MFCD00014999,MFCD00014999 |
InChI Key | GEBLOQXLELCEEO-UHFFFAOYSA-N |
Synonym | di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester |
PubChem CID | 21959072 |
IUPAC Name | ditert-butyl phosphite |
SMILES | CC(C)(C)O[P+](=O)OC(C)(C)C |
Description
Di-tert-butyl phosphite is used as a solvent, as an antioxidant, and as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13086-84-5 | |
193.20 | |
GEBLOQXLELCEEO-UHFFFAOYSA-N | |
21959072 | |
CC(C)(C)O[P+](=O)OC(C)(C)C |
C8H18O3P | |
MFCD00014999,MFCD00014999 | |
di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester | |
ditert-butyl phosphite |
Specifications
13086-84-5 | |
66°C to 68°C (0.5 mmHg) | |
1.42 | |
1 g | |
di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester | |
GEBLOQXLELCEEO-UHFFFAOYSA-N | |
ditert-butyl phosphite | |
21959072 | |
95% | |
Di-tert-butyl phosphite |
0.995 g/mL | |
C8H18O3P | |
MFCD00014999,MFCD00014999 | |
Air and moisture sensitive | |
Soluble in organic solvents | |
CC(C)(C)O[P+](=O)OC(C)(C)C | |
193.20 | |
194.21 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-996-8
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only