missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-Di-tert-butyl L-tartrate, 99%

Manufacturer:  Alfa Aesar™ H55860MD

 View more versions of this product

Catalog No. AAH55860MD



Specifications

Specifications

(+)-Di-tert-butyl L-tartrate
109°C (228°F)
90°C to 92°C
117384-45-9
MFCD00192000
+-di-tert-butyl l-tartrate
CC(C)(C)OC(=O)C(C(C(=O)OC(C)(C)C)O)O
262.302
262.3
99%
1.143 g/mL
White
250mg
C12H22O6
5026723
ITWOKJQQGHCDBL-HTQZYQBOSA-N
ditert-butyl (2R,3R)-2,3-dihydroxybutanedioate
9943166
Solid

Chemical Identifiers

117384-45-9
262.302
ITWOKJQQGHCDBL-HTQZYQBOSA-N
9943166
CC(C)(C)OC(=O)C(C(C(=O)OC(C)(C)C)O)O
C12H22O6
MFCD00192000
+-di-tert-butyl l-tartrate
ditert-butyl (2R,3R)-2,3-dihydroxybutanedioate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents