Please login to your online account to display your discounted pricing

Di-tert-butyl iminodicarboxylate, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC369660050

 View more versions of this product

Catalog No. AC369660050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Crystalline Powder
Formula Weight 217.27
Quantity 5g
Color White to beige
Fieser 01,210; 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268
Synonym (BOC)2NH
Chemical Name or Material Di-tert-butyl iminodicarboxylate
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H19NO4
Linear Formula (CH3)3COCO!2NH
CAS 51779-32-9

Specifications

Chemical Name or Material Di-tert-butyl iminodicarboxylate
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C10H19NO4
Linear Formula (CH3)3COCO!2NH
Formula Weight 217.27
Synonym (BOC)2NH