Please login to your online account to display your discounted pricing

Di-tert-butyl dicarbonate, 1M Solution in THF, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC426801000

 View more versions of this product

Catalog No. AC426801000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. May cause an allergic skin reaction. Fatal if inhaled. May cause drowsiness or dizziness. Suspected of causing cancer. May form explosive peroxides.
Formula Weight 218.25
Physical Form Liquid
Quantity 100mL
Color Clear colorless
Merck Index 15, 3038
Packaging AcroSeal™ Glass Bottle
Density 0.9130g/mL
Linear Formula O[CO2C(CH3)3]2
CAS 109-99-9
CAS 24424-99-5
Name Note 1M Solution in THF
Molecular Formula C10H18O5
Chemical Name or Material Di-tert-butyl dicarbonate
Health Hazard 1 Danger
MDL Number MFCD00008805

Specifications

Chemical Name or Material Di-tert-butyl dicarbonate
Name Note 1M Solution in THF
CAS 24424-99-5
CAS 109-99-9
Molecular Formula C10H18O5
Linear Formula O[CO2C(CH3)3]2
Formula Weight 218.25
MDL Number MFCD00008805
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
Highly flammable liquid and vapor. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. May cause an allergic skin reaction. Fatal if inhaled. May cause drowsiness or dizziness. Suspected of causing cancer. May form explosive peroxides.

SDS & Safety Documents