Learn More
Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 194671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-tert-butyl dicarbonate | |
24424-99-5 | |
White | |
56.0°C to 57.0°C (0.5 mmHg) | |
Authentic | |
Glass bottle | |
1.4075 to 1.4095 | |
MFCD00008805 | |
04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113 | |
15, 3038 | |
DYHSDKLCOJIUFX-UHFFFAOYSA-N | |
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate | |
90495 | |
218.25 | |
Low Melting Crystalline Solid |
99% | |
22.0°C to 24.0°C | |
1.0200g/mL | |
37°C | |
99% | |
C10H18O5 | |
O[CO2C(CH3)3]2 | |
100 g | |
1.02 | |
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate | |
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C | |
218.25 | |
CHEBI:48500 | |
99% |
Chemical Identifiers
24424-99-5 | |
218.25 | |
DYHSDKLCOJIUFX-UHFFFAOYSA-N | |
90495 | |
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate |
C10H18O5 | |
MFCD00008805 | |
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate | |
CHEBI:48500 | |
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti
GHS Signal Word: Danger
EINECSNumber : 246-240-1
RTECSNumber : HT0230000
TSCA : TSCA
RUO – Research Use Only