missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 366920050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
C-X Bond Formation (Halogen), Radical initiatorsSpecifications
Di-tert-butyl azodicarboxylate | |
Authentic | |
Glass bottle | |
(CH3)3CO2NNCO2C(CH3)3 | |
di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
QKSQWQOAUQFORH-VAWYXSNFSA-N | |
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate | |
6034084 | |
97% |
870-50-8 | |
96% min. (GC) | |
C10H18N2O4 | |
5g | |
Solubility in water: insoluble. Other solubilities: soluble in dichloromethane and other chlorinated,solvents | |
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C | |
230.26 | |
230.26 |
Chemical Identifiers
870-50-8 | |
230.26 | |
di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester | |
tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate |
C10H18N2O4 | |
QKSQWQOAUQFORH-VAWYXSNFSA-N | |
6034084 | |
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 212-796-9
RUO â Research Use Only