Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-sec-butylamine, (+/-) + meso, 98%

Chemical Identifiers

Chemical Name or Material Di-sec-butylamine
CAS 626-23-3
Name Note (+/-) + meso
Molecular Formula C8H19N
Formula Weight 129.25g/mol
MDL Number MFCD00009326
UN Number UN2733
Synonym Bis(2-methylpropyl)amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2569018

 
alfa aesar™
B25690-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2569030

 
alfa aesar™
B25690-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Amine-like
Melting Point −70°C
Boiling Point 134° to 136°C
Quantity 50g
Flash Point 24°C (75°F)
Chemical Name or Material Di-sec-butylamine
Name Note (+/-) + meso
CAS 626-23-3
Assay 98%
Molecular Formula C8H19N
Formula Weight 129.25g/mol
MDL Number MFCD00009326
UN Number UN2733
Synonym Bis(2-methylpropyl)amine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.