Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-p-tolyl sulfone, 98+%

$99.76 - $348.07

Chemical Identifiers

Chemical Name or Material Di-p-tolyl sulfone
CAS 599-66-6
Molecular Formula C14H14O2S
Formula Weight 246.32
MDL Number MFCD00041332
Synonym Bis(4-methylphenyl) sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1217606

  SDS
alfa aesar™
A1217606
5g Each for $99.76

AAA1217614

  SDS
alfa aesar™
A1217614
25g Each for $348.07
Description & Specifications

Specifications

Formula Weight 246.32
Assay ≥98%
TSCA TSCA
Synonym Bis(4-methylphenyl) sulfone
MDL Number MFCD00041332
Melting Point 156° to 161°C
CAS 599-66-6
Chemical Name or Material Di-p-tolyl sulfone
Molecular Formula C14H14O2S
Quantity 5g