missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(-)-Di-p-toluoyl-L-tartaric acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 117570250
Specifications
(-)-Di-p-toluoyl-L-tartaric acid | |
Glass bottle | |
-142 | |
White to Yellow | |
25g | |
96.0 | |
97% | |
[CH3C6H4CO2CH(CO2H)-]2 | |
--di-1,4-o-toluoyl-l-tartaric acid, --di-p-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r, di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, l-dtta | |
FOTRUJUPLHRVNU-QNBGGDODSA-N | |
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate | |
12297760 | |
Crystalline Powder |
Authentic | |
−142° (20°C c=10,C2H5OH) | |
0.5% max. | |
164°C to 170°C | |
32634-66-5 | |
100.0 | |
C20H18O8 | |
MFCD00064440 | |
Solubility in water: soluble. Other solubilities: soluble in acetone and ethanol | |
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O | |
386.36 | |
386.36 | |
97% |
Chemical Identifiers
32634-66-5 | |
386.36 | |
FOTRUJUPLHRVNU-QNBGGDODSA-N | |
12297760 | |
CC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)C)C(=O)O.O |
C20H18O8 | |
MFCD00064440 | |
--di-1,4-o-toluoyl-l-tartaric acid, --di-p-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r, di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, l-dtta | |
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate |
Safety and Handling
EINECSNumber : 251-131-7