missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1322322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-p-toluoyl-D-tartaric acid | |
32634-68-7 | |
MFCD00008552 | |
di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid | |
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O | |
386.36 | |
386.36 | |
100 g |
169°C to 171°C | |
C20H18O8 | |
3225584 | |
CMIBUZBMZCBCAT-HOTGVXAUSA-N | |
(2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid | |
263211 | |
98% | |
+139° (c=1 in Ethanol) |
Chemical Identifiers
32634-68-7 | |
386.36 | |
CMIBUZBMZCBCAT-HOTGVXAUSA-N | |
263211 |
C20H18O8 | |
MFCD00008552 | |
di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid | |
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O |
Safety and Handling
EINECSNumber : 251-132-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only