Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-p-toluoyl-D-tartaric acid, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1322306

 View more versions of this product

Catalog No. AAL1322306

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 386.36
Chemical Name or Material Di-p-toluoyl-D-tartaric acid
MDL Number MFCD00008552
Molecular Formula C20H18O8
CAS 32634-68-7
Assay 98%
Melting Point 169° to 171°C
TSCA TSCA
Beilstein 3225584
Optical Rotation +139° (c=1 in Ethanol)
Quantity 5g

Specifications

Chemical Name or Material Di-p-toluoyl-D-tartaric acid
CAS 32634-68-7
Molecular Formula C20H18O8
Formula Weight 386.36
MDL Number MFCD00008552
Beilstein 3225584