Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-O-acetyl-L-tartaric anhydride, 97%

$45.09 - $126.08

Chemical Identifiers

Chemical Name or Material Di-O-acetyl-L-tartaric anhydride
CAS 6283-74-5
Molecular Formula C8H8O7
Formula Weight 216.15
MDL Number MFCD00037918
Beilstein 87315
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1099809

  SDS
alfa aesar™
L1099809
10g Each for $45.09

AAL1099818

  SDS
alfa aesar™
L1099818
50g Each for $126.08
Description & Specifications

Specifications

Formula Weight 216.15
Chemical Name or Material Di-O-acetyl-L-tartaric anhydride
MDL Number MFCD00037918
CAS 6283-74-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Optical Rotation +60° (c=6 in Acetone)
Assay 97%
Molecular Formula C8H8O7
Beilstein 87315
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Melting Point 131° to 133°C
Quantity 10g

Applications
Di-O-acetyl-L-tartaric anhydride used as a chiral building block for proteomics research.

Solubility
Soluble in methanol and dichloromethane. Slightly soluble in water.

Notes
It is incompatible with strong oxidizing agents and strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C