missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-O-acetyl-L-tartaric anhydride, 97%

Manufacturer:  Alfa Aesar™ L1099818

 View more versions of this product

Catalog No. AAL1099818



Description

Description

Applications
Di-O-acetyl-L-tartaric anhydride used as a chiral building block for proteomics research.

Solubility
Soluble in methanol and dichloromethane. Slightly soluble in water.

Notes
It is incompatible with strong oxidizing agents and strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
Specifications

Specifications

Di-O-acetyl-L-tartaric anhydride
131°C to 133°C
6283-74-5
MFCD00037918
3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl diacetate, unii-d7xnf76o0k, +-diacetyl-l-tartaric anhydride, d7xnf76o0k, 2,5-furandione, 3,4-bis acetyloxy dihydro-, 3r,4r, 3r,4r-4-acetyloxy-2,5-dioxooxolan-3-yl acetate, di-o-acetyl-l-tartaric anhydride, o,o-diacetyl tartaric acid anhydride, acetic acid 4-acetoxy-2,5-dioxo-tetrahydro-furan-3-yl ester, 3r-trans-dihydro-2,5-dioxofuran-3,4-diyl diacetate
CC(=O)OC1C(C(=O)OC1=O)OC(=O)C
216.145
216.15
+60° (c=6 in Acetone)
50g
C8H8O7
87315
XAKITKDHDMPGPW-PHDIDXHHSA-N
[(3R,4R)-4-acetyloxy-2,5-dioxooxolan-3-yl] acetate
513914
97%

Chemical Identifiers

6283-74-5
216.145
XAKITKDHDMPGPW-PHDIDXHHSA-N
513914
CC(=O)OC1C(C(=O)OC1=O)OC(=O)C
C8H8O7
MFCD00037918
3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl diacetate, unii-d7xnf76o0k, +-diacetyl-l-tartaric anhydride, d7xnf76o0k, 2,5-furandione, 3,4-bis acetyloxy dihydro-, 3r,4r, 3r,4r-4-acetyloxy-2,5-dioxooxolan-3-yl acetate, di-o-acetyl-l-tartaric anhydride, o,o-diacetyl tartaric acid anhydride, acetic acid 4-acetoxy-2,5-dioxo-tetrahydro-furan-3-yl ester, 3r-trans-dihydro-2,5-dioxofuran-3,4-diyl diacetate
[(3R,4R)-4-acetyloxy-2,5-dioxooxolan-3-yl] acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents