Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-propylmalonic acid, 98%

$156.91 - $156.91

Chemical Identifiers

Chemical Name or Material Di-n-propylmalonic acid
CAS 1636-27-7
Molecular Formula C9H16O4
Formula Weight 188.23g/mol
MDL Number MFCD00051476
Beilstein 1775937
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0690406

 
alfa aesar™
L06904-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0690414

 
alfa aesar™
L06904-14
25g Each for $156.91
Description & Specifications

Specifications

Melting Point 154° to 156°C
Quantity 5g
Chemical Name or Material Di-n-propylmalonic acid
CAS 1636-27-7
Assay 98%
Molecular Formula C9H16O4
Formula Weight 188.23g/mol
MDL Number MFCD00051476
Beilstein 1775937
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.