missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-propyl sulfite, 99%

Manufacturer:  Alfa Aesar™ L1203314

 View more versions of this product

Catalog No. AAL1203314



Specifications

Specifications

Di-n-propyl sulfite
107°C to 109°C (42mmHg)
623-98-3
MFCD00059404
1703571
MAIQPVFXODAAIG-UHFFFAOYSA-N
dipropyl sulfite
136434
99%
110°C (230°F)
25g
C6H14O3S
UN3334
di-n-propyl sulfite, di-n-propyl sulphite, sulfurous acid, dipropyl ester, dipropyl sulphite, dipropylsulfite, di-n-propylsulfit, di-n-propyl sulfite, sulfurous acid dipropyl ester, sulphurous acid dipropyl ester, dipropyl ester of sulfurous acid
CCCOS(=O)OCCC
166.235
166.24

Chemical Identifiers

623-98-3
166.235
MAIQPVFXODAAIG-UHFFFAOYSA-N
136434
CCCOS(=O)OCCC
C6H14O3S
MFCD00059404
di-n-propyl sulfite, di-n-propyl sulphite, sulfurous acid, dipropyl ester, dipropyl sulphite, dipropylsulfite, di-n-propylsulfit, di-n-propyl sulfite, sulfurous acid dipropyl ester, sulphurous acid dipropyl ester, dipropyl ester of sulfurous acid
dipropyl sulfite
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.

Documents