Learn More
Di-n-propyl sulfide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2315536
Description
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
Solubility
Soluble in water 351mg/L @ 25°C.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Di-n-propyl sulfide | |
-102°C | |
141°C to 142°C | |
Stench | |
1.4487 | |
500 g | |
1719002 | |
propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane | |
ZERULLAPCVRMCO-UHFFFAOYSA-N | |
1-propylsulfanylpropane | |
8118 | |
≥98% |
111-47-7 | |
0.838 | |
28°C (82°F) | |
C6H14S | |
MFCD00009379 | |
UN1993 | |
14,7868 | |
Soluble in water 351mg/L @ 25°C. | |
CCCSCCC | |
118.24 | |
118.24 |
Chemical Identifiers
111-47-7 | |
118.24 | |
ZERULLAPCVRMCO-UHFFFAOYSA-N | |
8118 | |
CCCSCCC |
C6H14S | |
MFCD00009379 | |
propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane | |
1-propylsulfanylpropane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 203-873-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only