Learn More
Di-n-propyl phthalate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2405618
Description
Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.
Solubility
Soluble in chloroform, methanol.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Di-n-propyl phthalate | |
1.082 | |
>110°C (230°F) | |
1.497 | |
50 g | |
2332522 | |
Soluble in chloroform,methanol. | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC | |
250.294 | |
CHEBI:60069 | |
98% |
131-16-8 | |
168°C to 170°C (3 mmHg) | |
C14H18O4 | |
MFCD00009371 | |
UN3082 | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
dipropyl benzene-1,2-dicarboxylate | |
8559 | |
250.3 |
Chemical Identifiers
131-16-8 | |
250.294 | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
8559 | |
dipropyl benzene-1,2-dicarboxylate |
C14H18O4 | |
MFCD00009371 | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
CHEBI:60069 | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P280-P308+P313-P501c
H351-H361
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 205-015-8
RTECSNumber : TI1940000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only