Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-pentylphenylphosphine, 97%

$68.50 - $266.30

Chemical Identifiers

Chemical Name or Material Di-n-pentylphenylphosphine
CAS 71501-08-1
Molecular Formula [CH3(CH2)4]2PC6H5
Formula Weight 250.35
MDL Number MFCD00015273
Synonym Di-n-pentylphenylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1832503

 
alfa aesar™
A1832503
1g Each for $68.50

AAA1832506

 
alfa aesar™
A1832506
5g Each for $266.30
Description & Specifications

Specifications

Formula Weight 250.35
Chemical Name or Material Di-n-pentylphenylphosphine
Assay 97%
Molecular Formula [CH3(CH2)4]2PC6H5
MDL Number MFCD00015273
CAS 71501-08-1
Synonym Di-n-pentylphenylphosphine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 209° to 211°C (50mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.