missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-octyltin oxide

Manufacturer:  Alfa Aesar™ 7119336

 View more versions of this product

Catalog No. AA7119336



Specifications

Specifications

Di-n-octyltin oxide
870-08-6
White
MFCD00013839
LQRUPWUPINJLMU-UHFFFAOYSA-N
dioctyl(oxo)tin
93563
Powder
245°C to 248°C (decomposition)
500g
C16H34OSn
di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
CCCCCCCC[Sn](=O)CCCCCCCC
361.157
361.14

Chemical Identifiers

870-08-6
361.157
LQRUPWUPINJLMU-UHFFFAOYSA-N
93563
CCCCCCCC[Sn](=O)CCCCCCCC
C16H34OSn
MFCD00013839
di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
dioctyl(oxo)tin
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WH7620000

TSCA : TSCA

SDS Documents