missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-n-octylamine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 117081000
Specifications
Di-n-octylamine | |
>110°C | |
Glass bottle | |
0.79 | |
Colorless to Yellow | |
100g | |
96% min. (GC) | |
[CH3(CH2)7]2NH | |
04, 196 | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
N-octyloctan-1-amine | |
3094 | |
Liquid |
0.7900g/mL | |
Authentic | |
1.4412 to 1.4432 | |
297°C to 298°C | |
14°C to 15°C | |
1120-48-5 | |
C16H35N | |
MFCD00009557 | |
1-octanamine, n-octyl, di n-octyl amine, di-n-octylamine, di-normal-octylamine, dioctyl-amine, dioctylamine, n,n-dioctylamine, n-n-octyl-n-octylamine, unii-a7hm3062rm | |
CCCCCCCCNCCCCCCCC | |
241.46 | |
241.46 | |
97% |
Chemical Identifiers
1120-48-5 | |
241.46 | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
3094 | |
CCCCCCCCNCCCCCCCC |
C16H35N | |
MFCD00009557 | |
1-octanamine, n-octyl, di n-octyl amine, di-n-octylamine, di-normal-octylamine, dioctyl-amine, dioctylamine, n,n-dioctylamine, n-n-octyl-n-octylamine, unii-a7hm3062rm | |
N-octyloctan-1-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 214-311-6