Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-nonyl sulfide, 98%

$43.84 - $149.92

Chemical Identifiers

Chemical Name or Material Di-n-nonyl sulfide
CAS 929-98-6
Molecular Formula C18H38S
Linear Formula CH3(CH2)8S(CH2)8CH3
Formula Weight 286.56
MDL Number MFCD00027342
Beilstein 1755874
Synonym n-Nonyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1063814

  SDS
alfa aesar™
L1063814
25g Each for $149.92

AAL1063806

  SDS
alfa aesar™
L1063806
5g Each for $43.84
Description & Specifications

Specifications

Formula Weight 286.56
Chemical Name or Material Di-n-nonyl sulfide
MDL Number MFCD00027342
CAS 929-98-6
Molecular Formula C18H38S
Synonym n-Nonyl sulfide
Quantity 25g
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)8S(CH2)8CH3
Beilstein 1755874
Odor Unpleasant
Melting Point 15°C
Boiling Point 165° to 167°C (3mmHg)
Assay 98%