missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-nonyl sulfide, 98%

Manufacturer:  Alfa Aesar™ L1063806

 View more versions of this product

Catalog No. AAL1063806



Specifications

Specifications

Di-n-nonyl sulfide
Unpleasant
15°C
929-98-6
CH3(CH2)8S(CH2)8CH3
1755874
KMKSVAGOBVUFRO-UHFFFAOYSA-N
1-nonylsulfanylnonane
525006
98%
>110°C (230°F)
165°C to 167°C (3mmHg)
5g
C18H38S
MFCD00027342
dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
CCCCCCCCCSCCCCCCCCC
286.562
286.56

Chemical Identifiers

929-98-6
286.562
KMKSVAGOBVUFRO-UHFFFAOYSA-N
525006
CCCCCCCCCSCCCCCCCCC
C18H38S
MFCD00027342
dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
1-nonylsulfanylnonane
Documents