missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-n-hexyl sulfide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2232514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-n-hexyl sulfide | |
0.849 | |
116°C (241°F) | |
C12H26S | |
MFCD00009528 | |
UN3334 | |
hexyl sulfide, hexane, 1,1'-thiobis, dihexyl sulfide, di-n-hexyl sulfide, 7-thiatridecane, dihexyl sulphide, n-hexyl sulfide, di-n-hexylsulfide, hexylthiohexane, normal-hexyl sulfide | |
CCCCCCSCCCCCC | |
202.40 | |
202.4 |
6294-31-1 | |
260°C | |
Stench | |
1.4587 | |
25 g | |
1738333 | |
LHNRHYOMDUJLLM-UHFFFAOYSA-N | |
1-hexylsulfanylhexane | |
80517 | |
97% |
Chemical Identifiers
6294-31-1 | |
202.40 | |
LHNRHYOMDUJLLM-UHFFFAOYSA-N | |
80517 | |
CCCCCCSCCCCCC |
C12H26S | |
MFCD00009528 | |
hexyl sulfide, hexane, 1,1'-thiobis, dihexyl sulfide, di-n-hexyl sulfide, 7-thiatridecane, dihexyl sulphide, n-hexyl sulfide, di-n-hexylsulfide, hexylthiohexane, normal-hexyl sulfide | |
1-hexylsulfanylhexane |
Safety and Handling
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber : 228-556-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only