missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1259309
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.
Solubility
Soluble in water (0.24 mg/L).
Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.
Specifications
Di-n-hexyl phthalate | |
1.01 | |
C20H30O4 | |
MFCD00043720 | |
UN3082 | |
Soluble in water (0.24mg/L). | |
CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC | |
334.46 | |
CHEBI:34678 | |
97% |
84-75-3 | |
185°C to 187°C (0.5 mmHg) | |
1.487 | |
10 g | |
dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate | |
KCXZNSGUUQJJTR-UHFFFAOYSA-N | |
dihexyl benzene-1,2-dicarboxylate | |
6786 | |
334.46 |
Chemical Identifiers
84-75-3 | |
334.46 | |
KCXZNSGUUQJJTR-UHFFFAOYSA-N | |
6786 | |
dihexyl benzene-1,2-dicarboxylate |
C20H30O4 | |
MFCD00043720 | |
dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate | |
CHEBI:34678 | |
CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360FD
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 201-559-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only