missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-n-hexyl disulfide, tech. 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2256806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-n-hexyl disulfide | |
0.919 | |
Unpleasant | |
MFCD00027300 | |
hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide | |
CCCCCCSSCCCCCC | |
234.46 | |
234.47 | |
Technical |
10496-15-8 | |
>110°C (230°F) | |
C12H26S2 | |
5 g | |
GJPDBURPGLWRPW-UHFFFAOYSA-N | |
1-(hexyldisulfanyl)hexane | |
82675 | |
85% |
Chemical Identifiers
10496-15-8 | |
234.46 | |
GJPDBURPGLWRPW-UHFFFAOYSA-N | |
82675 | |
CCCCCCSSCCCCCC |
C12H26S2 | |
MFCD00027300 | |
hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide | |
1-(hexyldisulfanyl)hexane |
Safety and Handling
EINECSNumber : 234-022-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only