missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-heptyl phthalate, 98+%

Manufacturer:  Alfa Aesar™ A1546630

 View more versions of this product

Catalog No. AAA1546630



Description

Description

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.
Specifications

Specifications

Di-n-heptyl phthalate
360°C
3648-21-3
MFCD00043680
diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC
362.51
CHEBI:34677
≥98%
213°C (415°F)
250g
C22H34O4
2336623
JQCXWCOOWVGKMT-UHFFFAOYSA-N
diheptyl benzene-1,2-dicarboxylate
19284
362.51

Chemical Identifiers

3648-21-3
362.51
JQCXWCOOWVGKMT-UHFFFAOYSA-N
19284
diheptyl benzene-1,2-dicarboxylate
C22H34O4
MFCD00043680
diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
CHEBI:34677
CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TI1090000

TSCA : TSCA

Documents