missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyltin oxide

Manufacturer:  Alfa Aesar™ 14117A1

 View more versions of this product

Catalog No. AA14117A1



Specifications

Specifications

Di-n-butyltin oxide
818-08-6
Odorless
White
[CH3(CH2)3]2SnO
UN3146
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
CCCC[Sn](=O)CCCC
248.941
248.92
>300°C
1kg
1.58g/mL
C8H18OSn
MFCD00001992
4126243
JGFBRKRYDCGYKD-UHFFFAOYSA-N
dibutyl(oxo)tin
61221
Powder

Chemical Identifiers

818-08-6
248.941
JGFBRKRYDCGYKD-UHFFFAOYSA-N
61221
CCCC[Sn](=O)CCCC
C8H18OSn
MFCD00001992
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
dibutyl(oxo)tin
Safety and Handling

Safety and Handling

GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH7175000

TSCA : TSCA

Documents