missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyltin dilaurate, 95%

Manufacturer:  Alfa Aesar™ 7113036

 View more versions of this product

Catalog No. AA7113036



Specifications

Specifications

Di-n-butyltin dilaurate
<10°C
500g
1.066g/mL
1.4730
C32H64O4Sn
MFCD00008963
14,3038
Soluble in benzene and ether; Insoluble in water and methanol
Liquid
>204°C (12mmHg)
77-58-7
235°C (455°F)
Yellow
95%
[CH3(CH2)3]2Sn[O2C(CH2)10CH3]2
UN2788
Di-n-butyltin didodecanoate
631.55

Chemical Identifiers

77-58-7
C32H64O4Sn
MFCD00008963
Di-n-butyltin didodecanoate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H360-H341-H314-H318-H317
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH7000000

TSCA : TSCA

SDS Documents