missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyltin dichloride, 96%

Organotin intermediate

Manufacturer:  Alfa Aesar™ 1411609

 View more versions of this product

Catalog No. AA1411609



Specifications

Specifications

Di-n-butyltin dichloride
39°C to 41°C
10g
Acrid
White
Moisture Sensitive
[CH3(CH2)3]2SnCl2
UN2928
Insoluble in cold water; Hydrolyzed by hot water; Soluble in organic solvents
CCCC[Sn](CCCC)(Cl)Cl
303.842
303.83
96%
135°C (10mmHg)
683-18-1
168° (334°F)
1.36g/mL
1.4991
C8H18Cl2Sn
MFCD00000518
dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
RJGHQTVXGKYATR-UHFFFAOYSA-L
dibutyl(dichloro)stannane
12688
Crystalline

Chemical Identifiers

683-18-1
303.842
RJGHQTVXGKYATR-UHFFFAOYSA-L
12688
CCCC[Sn](CCCC)(Cl)Cl
C8H18Cl2Sn
MFCD00000518
dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
dibutyl(dichloro)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H360-H372-H341-H314-H312
Toxic if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : WH7100000

TSCA : TSCA

Documents