missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyltin diacetate, 95%

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions

Manufacturer:  Alfa Aesar™ 1859518

 View more versions of this product

Catalog No. AA1859518



Specifications

Specifications

Di-n-butyltin diacetate
12°C
50g
Acidic
Colorless to Yellow
[OOCCH3]2Sn[(CH2)3CH3]2
UN2788
Soluble in water and most organic solvents
Liquid
142°C to 145°C (10mmHg)
1067-33-0
143° (290°F)
1.32g/mL
1.4700
MFCD00008697
Diacetoxydi-n-butyltin
351.01
95%

Chemical Identifiers

1067-33-0
MFCD00008697
Diacetoxydi-n-butyltin
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH6880000

TSCA : TSCA

SDS Documents