missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions

Supplier:  Thermo Scientific Chemicals 018595A1

 View more versions of this product

Catalog No. AA18595A1



Description

Description

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-n-butyltin diacetate
12°C
1.32 g/mL
143°C (290°F)
C12H26O4Sn
[OOCCH3]2Sn[(CH2)3CH3]2
1 kg
Diacetoxydi-n-butyltin
LDZDQDDROJXUBO-UHFFFAOYSA-N
353.05
95%
1067-33-0
Colorless to Yellow
142°C to 145°C (10 mmHg)
Acidic
1.47
MFCD00008697
UN2788
Soluble in water and most organic solvents; Miscible with most organic solvents.
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
351.01
Liquid

Chemical Identifiers

1067-33-0
353.05
LDZDQDDROJXUBO-UHFFFAOYSA-N
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
C12H26O4Sn
MFCD00008697
Diacetoxydi-n-butyltin
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H301-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 213-928-8

RTECSNumber : WH6880000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only