Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butylgermanium dichloride, 97%

Chemical Identifiers

Chemical Name or Material Di-n-butylgermanium dichloride
CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Linear Formula [CH3(CH2)3]2GeCl2
Formula Weight 257.73
MDL Number MFCD00013590
UN Number UN2922
Synonym Di-n-butyldichlorogermane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1713814

 
alfa aesar™
A1713814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.73
Chemical Name or Material Di-n-butylgermanium dichloride
MDL Number MFCD00013590
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 241° to 243°C
TSCA TSCA
Assay 97%
Molecular Formula C8H18Cl2Ge
Linear Formula [CH3(CH2)3]2GeCl2
CAS 4593-81-1
Quantity 25g
UN Number UN2922
Synonym Di-n-butyldichlorogermane
Flash Point 107°C (225°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number LY5020000
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.