Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butylammonium tetrafluoroborate, 98%

$31.09 - $94.97

Chemical Identifiers

Chemical Name or Material Di-n-butylammonium tetrafluoroborate
CAS 12107-76-5
Molecular Formula C8H20BF4N
Linear Formula (C4H9)2NH2BF4
Formula Weight 217.06
MDL Number MFCD00034899
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2025122

 
alfa aesar™
B2025122
100g Each for $94.97

AAB2025114

 
alfa aesar™
B2025114
25g Each for $31.09
Description & Specifications

Specifications

Formula Weight 217.06
Chemical Name or Material Di-n-butylammonium tetrafluoroborate
TSCA TSCA
Quantity 100g
Assay 98%
Molecular Formula C8H20BF4N
Linear Formula (C4H9)2NH2BF4
MDL Number MFCD00034899
CAS 12107-76-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.