Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyl phosphate, 96%

Chemical Identifiers

Chemical Name or Material Di-n-butyl phosphate
CAS 107-66-4
Molecular Formula (CH3(CH2)3O)2P(O)OH
Formula Weight 210.21
MDL Number MFCD00015245
UN Number UN1718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43371AK

  SDS
alfa aesar™
43371AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43371K2

  SDS
alfa aesar™
43371K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.21
RTECS Number TB9605000
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 250°C
Melting Point −13°C
Molecular Formula (CH3(CH2)3O)2P(O)OH
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYL ACID PHOSPHATE
Chemical Name or Material Di-n-butyl phosphate
UN Number UN1718
Refractive Index 1.428
pH 1.0
MDL Number MFCD00015245
Flash Point 157°C (315°F)
Physical Form Liquid
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Density 1.058g/mL
Odor Odorless
CAS 107-66-4
Assay 96%
Quantity 250mL
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.