Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyl N,N-diethylcarbamoylphosphonate, 94%


Quantity 25g

Manufacturer: alfa aesar™  L0448014

 View more versions of this product

Catalog No. AAL0448014

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 293.34
Chemical Name or Material Di-n-butyl N,N-diethylcarbamoylphosphonate
MDL Number MFCD00039952
CAS 7439-69-2
Assay 94%
Beilstein 1796281
Quantity 25g
Boiling Point 132° to 134°C (1mmHg)
Molecular Formula C13H28NO4P

Specifications

Chemical Name or Material Di-n-butyl N,N-diethylcarbamoylphosphonate
CAS 7439-69-2
Molecular Formula C13H28NO4P
Formula Weight 293.34
MDL Number MFCD00039952
Beilstein 1796281