Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-n-butyl N,N-diethylcarbamoylphosphonate, 94%

$81.22 - $302.86

Chemical Identifiers

Chemical Name or Material Di-n-butyl N,N-diethylcarbamoylphosphonate
CAS 7439-69-2
Molecular Formula C13H28NO4P
Formula Weight 293.34
MDL Number MFCD00039952
Beilstein 1796281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0448006

  SDS
alfa aesar™
L0448006
5g Each for $81.22

AAL0448014

  SDS
alfa aesar™
L0448014
25g Each for $302.86
Description & Specifications

Specifications

Formula Weight 293.34
CAS 7439-69-2
Chemical Name or Material Di-n-butyl N,N-diethylcarbamoylphosphonate
MDL Number MFCD00039952
Molecular Formula C13H28NO4P
Assay 94%
Beilstein 1796281
Quantity 5g
Boiling Point 132° to 134°C (1mmHg)