Alfa Aesar™ Di-n-butyl methylphosphonate
Manufacturer:  Alfa Aesar™ 3046704
 View more versions of this product
Catalog No. AA3046704
Specifications
Specifications
Di-n-butyl methylphosphonate
2404-73-1
MFCD00015242
dibutyl methylphosphonate, dibutyl methanephosphonate, di-n-butyl methylphosphonate, dibutylmethylphosphonate, di-n-butyl methanephosphonate, methylphosphonic acid dibutyl ester, methylphosphonic acid, dibutyl ester, 1-butoxy methyl phosphoryl oxybutane, phosphonic acid, methyl-, dibutyl ester, phosphonic acid, methyl-, o,o-dibutyl ester
CCCCOP(=O)(C)OCCCC
208.238
208.24
2g
C9H21O3P
UN3272
NPNBLTJGRBYCJB-UHFFFAOYSA-N
1-[butoxy(methyl)phosphoryl]oxybutane
16994
Liquid
Chemical Identifiers
2404-73-1
208.238
NPNBLTJGRBYCJB-UHFFFAOYSA-N
16994
CCCCOP(=O)(C)OCCCC
C9H21O3P
MFCD00015242
dibutyl methylphosphonate, dibutyl methanephosphonate, di-n-butyl methylphosphonate, dibutylmethylphosphonate, di-n-butyl methanephosphonate, methylphosphonic acid dibutyl ester, methylphosphonic acid, dibutyl ester, 1-butoxy methyl phosphoryl oxybutane, phosphonic acid, methyl-, dibutyl ester, phosphonic acid, methyl-, o,o-dibutyl ester
1-[butoxy(methyl)phosphoryl]oxybutane
Safety and Handling
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
SDS Documents