missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-n-butyl ether, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A163050F

 View more versions of this product

Catalog No. AAA163050F



Description

Description

Di-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.

Solubility
Miscible with alcohol, ether and acetone. Immiscible with water.

Notes
Air and light sensitive. Incompatible with oxidizing agents and strong acids.
Specifications

Specifications

Di-n-butyl ether
142-96-1
141°C to 143°C
25°C (77°F)
C8H18O
UN1149
14,1569
Miscible with alcohol,ether and acetone. Immiscible with water.
CCCCOCCCC
130.23
130.23
-95°C
0.764
2500 mL
1.399
MFCD00009461
1732752
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
DURPTKYDGMDSBL-UHFFFAOYSA-N
1-butoxybutane
8909
99%

Chemical Identifiers

142-96-1
130.23
DURPTKYDGMDSBL-UHFFFAOYSA-N
8909
CCCCOCCCC
C8H18O
MFCD00009461
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
1-butoxybutane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIBUTYL ETHERS

EINECSNumber : 205-575-3

RTECSNumber : EK5425000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only