missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-mu-chlorobis(norbornadiene)dirhodium(I), Rh 44% min

Manufacturer:  Alfa Aesar™ 1046702

 View more versions of this product

Catalog No. AA1046702



Specifications

Specifications

Di-mu-chlorobis(norbornadiene)dirhodium(I)
0.5g
C14H16Cl2Rh
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer
HCZNBZKRJTYNCE-UHFFFAOYSA-L
bicyclo[2.2.1]hepta-2,5-diene;rhodium(2+);dichloride
114600
Powder
Odorless
12257-42-0
MFCD00198060
Rh 44% min.
C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Cl-].[Cl-].[Rh+2]
358.087
461.01

Chemical Identifiers

12257-42-0
358.087
HCZNBZKRJTYNCE-UHFFFAOYSA-L
114600
C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Cl-].[Cl-].[Rh+2]
C14H16Cl2Rh
MFCD00198060
bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer
bicyclo[2.2.1]hepta-2,5-diene;rhodium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents