missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-mu-bromobis(tri-tert-butylphosphine)dipalladium(I)

Manufacturer:  Alfa Aesar™ 4444604

 View more versions of this product

Catalog No. AA4444604



Description

Description

  • Coupling reactions
  • Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides
  • Especially active in difficult aminations
Specifications

Specifications

Di-mu-bromobis (tri-tert-butylphosphine)dipalladium (I)
Green
185812-86-6
[Pd(C4H9)3PBr]2
acmc-1c5o0
WIADNXKBAGKNOT-UHFFFAOYSA-M
palladium;tritert-butylphosphane;bromide
71317307
Powder
Air sensitive
2g
C12H27BrPPd-
MFCD04114019
Soluble in benzene and tolueneSoluble in tetrahydrofuran,toluene,dichloromethane and chloroform; Insoluble in alcohols and water.
CC(C)(C)P(C(C)(C)C)C(C)(C)C.[Br-].[Pd]
388.646
777.29

Chemical Identifiers

185812-86-6
388.646
WIADNXKBAGKNOT-UHFFFAOYSA-M
71317307
CC(C)(C)P(C(C)(C)C)C(C)(C)C.[Br-].[Pd]
C12H27BrPPd-
MFCD04114019
acmc-1c5o0
palladium;tritert-butylphosphane;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

Documents