Please login to your online account to display your discounted pricing

Di-μ-iodobis(p-cymene)iodoruthenium(II) 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382570010

 View more versions of this product

Catalog No. AC382570010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 978.18
Physical Form Crystalline Powder
Quantity 1g
Color Dark bordeaux
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Molecular Formula C20H28I4Ru2
Chemical Name or Material Di-μ-iodobis(p-cymene)iodoruthenium(II)
CAS 90614-07-6
Elemental Analysis 96.0% min.
MDL Number MFCD08064027
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Di-μ-iodobis(p-cymene)iodoruthenium(II)
Assay Percent Range 97%
Molecular Formula C20H28I4Ru2
Formula Weight 978.18
MDL Number MFCD08064027
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

SDS & Safety Documents