Please login to your online account to display your discounted pricing

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  348980050

 View more versions of this product

Catalog No. AC348980050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 464.42
Quantity 5g
Color Dark purple
Chemical Name or Material Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12H34Cl2Cu2N4O2
MDL Number MFCD01321163
CAS 30698-64-7
Synonym Cu-TMEDA catalyst
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride
CAS 30698-64-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H34Cl2Cu2N4O2
Formula Weight 464.42
MDL Number MFCD01321163
Synonym Cu-TMEDA catalyst