missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

di-Ammonium Hydrogen Phosphate, MilliporeSigma™

For analysis EMSURE™ ACS, Reag. Ph Eur

Manufacturer:  MilliporeSigma 1012070500

 View more versions of this product

Catalog No. M1012070500



Specifications

Specifications

di-Ammonium hydrogen phosphate
H9N2O4P
Soluble in water: 690g/L at 20°C
[NH4+].[NH4+].OP(=O)([O-])[O-]
132.056
CHEBI:63051
1.619g/cm3 at 20°C
Plastic Bottle
500g
7783-28-0
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diazanium;hydrogen phosphate
24540
132.05
Stinging
ACS, Reag. Ph Eur

Chemical Identifiers

7783-28-0
132.056
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
CHEBI:63051
[NH4+].[NH4+].OP(=O)([O-])[O-]
H9N2O4P
MNNHAPBLZZVQHP-UHFFFAOYSA-N
24540
diazanium;hydrogen phosphate
Safety and Handling

Safety and Handling

EINECSNumber : 231-987-8

Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry

Documents
Promotions

Promotions