missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-2-thienyl ketoxime, 97%

$161.55 - $161.55

Chemical Identifiers

CAS 10558-44-8
Molecular Formula C9H7NOS2
Formula Weight 209.29
MDL Number MFCD00209632
Synonym Bis-2-thienyl ketoxime; Di(2-thienyl)methanone oxime
Chemical Name or Material Di-2-thienyl ketoxime
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1818306 SDS Alfa Aesar™
A1818306
5g Each for $161.55
AAA1818303 SDS Alfa Aesar™
A1818303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10558-44-8
209.29
Bis-2-thienyl ketoxime; Di(2-thienyl)methanone oxime
C9H7NOS2
MFCD00209632
Di-2-thienyl ketoxime

Specifications

130° to 134°C
10558-44-8
209.29
Bis-2-thienyl ketoxime; Di(2-thienyl)methanone oxime
97%
5g
C9H7NOS2
MFCD00209632
Di-2-thienyl ketoxime