Please login to your online account to display your discounted pricing

Alfa Aesar™ Di(2-thienyl)-1,1,2,2-tetramethyldisilane, 97%

$131.99 - $390.26

Chemical Identifiers

Chemical Name or Material Di(2-thienyl)-1,1,2,2-tetramethyldisilane
CAS 124733-24-0
Molecular Formula C12H18S2Si2
Formula Weight 282.57
MDL Number MFCD09266191
Synonym Bis(2-thienyl)-1,1,2,2-tetramethyldisilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2893803

  SDS
alfa aesar™
H2893803
1g Each for $131.99

AAH2893806

  SDS
alfa aesar™
H2893806
5g Each for $390.26
Description & Specifications

Specifications

Formula Weight 282.57
Quantity 1g
Assay 97%
MDL Number MFCD09266191
CAS 124733-24-0
Synonym Bis(2-thienyl)-1,1,2,2-tetramethyldisilane
Molecular Formula C12H18S2Si2
Melting Point 42° to 44°C
Chemical Name or Material Di(2-thienyl)-1,1,2,2-tetramethyldisilane