Learn More
Di-2-pyridyl thionocarbonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2245114
Description
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.
Solubility
Soluble in water 9280 g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Di-2-pyridyl thionocarbonate | |
96°C to 99°C | |
MFCD00074870 | |
Moisture sensitive | |
Soluble in water 9280g/L). | |
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2 | |
232.257 | |
232.26 |
96989-50-3 | |
C11H8N2O2S | |
25 g | |
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate | |
IKYOVSVBLHGFMA-UHFFFAOYSA-N | |
dipyridin-2-yloxymethanethione | |
719784 | |
98% |
Chemical Identifiers
96989-50-3 | |
232.257 | |
IKYOVSVBLHGFMA-UHFFFAOYSA-N | |
719784 | |
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2 |
C11H8N2O2S | |
MFCD00074870 | |
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate | |
dipyridin-2-yloxymethanethione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only