Learn More
Di(2-pyridyl) ketone, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific H5532214
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applicationsdi-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a ″star″ topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.
Solubility
Low solubility in water.
Notes
Desiccate at room temperature. Keep away from oxidizing agents.
Specifications
Di(2-pyridyl) ketone | |
52°C to 55°C | |
C11H8N2O | |
126598 | |
25g | |
QPOWUYJWCJRLEE-UHFFFAOYSA-N | |
dipyridin-2-ylmethanone | |
88065 | |
99% |
19437-26-4 | |
110°C (230°F) | |
MFCD00006288 | |
2,2'-carbonyldipyridine, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, bis 2-pyridyl ketone, di 2-pyridyl ketone, di pyridin-2-yl methanone, di-2-pyridyl ketone, methanone, di-2-pyridinyl, pyridyl ketone | |
Low solubility in water. | |
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2 | |
184.198 | |
184.2 |
Chemical Identifiers
19437-26-4 | |
184.198 | |
QPOWUYJWCJRLEE-UHFFFAOYSA-N | |
88065 | |
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2 |
C11H8N2O | |
MFCD00006288 | |
2,2'-carbonyldipyridine, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, bis 2-pyridyl ketone, di 2-pyridyl ketone, di pyridin-2-yl methanone, di-2-pyridyl ketone, methanone, di-2-pyridinyl, pyridyl ketone | |
dipyridin-2-ylmethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-067-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only