missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Di(2-pyridyl) ketone, 99%

Manufacturer:  Alfa Aesar™ H5532206

 View more versions of this product

Catalog No. AAH5532206



Description

Description

Applications
di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a ″star″ topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

Solubility
Low solubility in water.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

Di(2-pyridyl) ketone
52°C to 55°C
19437-26-4
MFCD00006288
2,2'-carbonyldipyridine
QPOWUYJWCJRLEE-UHFFFAOYSA-N
dipyridin-2-ylmethanone
88065
99%
110°C (230°F)
5g
C11H8N2O
126598
Low solubility in water.
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2
184.198
184.2

Chemical Identifiers

19437-26-4
184.198
QPOWUYJWCJRLEE-UHFFFAOYSA-N
88065
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2
C11H8N2O
MFCD00006288
2,2'-carbonyldipyridine
dipyridin-2-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 243-067-3

TSCA : No

Recommended Storage : Ambient temperatures

Documents