missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-2-pyridyl ketone, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC155670250

 View more versions of this product

Catalog No. AC155670250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Di-2-pyridyl ketone
Glass bottle
49°C to 54°C
98%
98%
(C5H4N)CO(C5H4N)
24, IV, 591
QPOWUYJWCJRLEE-UHFFFAOYSA-N
dipyridin-2-ylmethanone
88065
Liquid
Authentic
Colorless to Yellow
25g
19437-26-4
C11H8N2O
MFCD00006288
di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2
184.198
184.2
≥97.5% (GC)

Chemical Identifiers

19437-26-4
184.198
QPOWUYJWCJRLEE-UHFFFAOYSA-N
88065
C1=CC=NC(=C1)C(=O)C2=CC=CC=N2
C11H8N2O
MFCD00006288
di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
dipyridin-2-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents